siRNA Details
| Gene Symbol: Pds5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081321.1 | NP_001074790.1 | 23 | 2738 | ||
| GenBank mRNA | AK172992.1 | - | 22 | 4435 | ||
| BC021408.1 | - | 10 | 1121 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pds5a| Gene Symbol: Pds5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081321.1 | NP_001074790.1 | 23 | 2738 | ||
| GenBank mRNA | AK172992.1 | - | 22 | 4435 | ||
| BC021408.1 | - | 10 | 1121 | |||