siRNA Details
| Gene Symbol: Gpr160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134385.2 | NP_001127857.1 | 4 | 1029 | ||
| NM_001134386.2 | NP_001127858.1 | 3 | 512 | |||
| NM_001286994.1 | NP_001273923.1 | 4 | 1038 | |||
| NM_001286995.1 | NP_001273924.1 | 3 | 741 | |||
| NM_001286996.1 | NP_001273925.1 | 4 | 612 | |||
| NM_027965.2 | NP_082241.1 | 4 | 841 | |||
| GenBank mRNA | AK006330.1 | - | 4 | 802 | ||
| AK154620.1 | - | 3 | 514 | |||
| AK154784.1 | - | 4 | 996 | |||
| BC116649.1 | - | 4 | 964 | 40088354 | ||

Gpr160
Genomic Map