siRNA Details
| Gene Symbol: Jakmip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028708.2 | NP_082984.1 | 1582 | |||
| GenBank mRNA | AK220482.1 | - | 1562 | |||
| BC119557.1 | - | 1256 | ||||
| BC119558.1 | - | 1247 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jakmip3| Gene Symbol: Jakmip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028708.2 | NP_082984.1 | 1582 | |||
| GenBank mRNA | AK220482.1 | - | 1562 | |||
| BC119557.1 | - | 1256 | ||||
| BC119558.1 | - | 1247 | ||||