siRNA Details
| Gene Symbol: Rtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030112.2 | NP_084388.2 | 9 | 1243 | ||
| GenBank mRNA | AK135633.1 | - | 3 | 311 | ||
| BC117848.1 | - | 9 | 1095 | 40093717 | ||
| BC117849.1 | - | 9 | 1095 | 40093724 | ||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtf1| Gene Symbol: Rtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030112.2 | NP_084388.2 | 9 | 1243 | ||
| GenBank mRNA | AK135633.1 | - | 3 | 311 | ||
| BC117848.1 | - | 9 | 1095 | 40093717 | ||
| BC117849.1 | - | 9 | 1095 | 40093724 | ||
Genomic Map