This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SPCS3 SIRNA Assays ›
Gene Symbol:
Spcs3
Gene Name:
signal peptidase complex subunit 3 homolog (S. cerevisiae)
Gene Aliases:
1810011E08Rik
Chromosome Location:
Chr.8: 54520433 - 54530739 on Build GRCm38
UniGene ID:
Mm.86437
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS293723
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Spcs3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029701.1 NP_083977.1 1 164
GenBank mRNA AK007432.1 - 1 143
AK028314.1 - 1 126
AK048051.1 - 1 150
BC019390.1 - 1 103
BC054817.1 - 1 129 5375997
BY012914.1 - 1 164

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_021928, AK026302, AY359044, BC053883, BC047290, and others.

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fh6lz:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline