siRNA Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF376726.1 | - | 12, 13 | 1317 | ||
| AK019012.1 | - | 3, 4 | 291 | |||
| AK047364.1 | - | 12, 13 | 1293 | |||
| AK082797.1 | - | 12, 13 | 1342 | |||
| AK159313.1 | - | 12, 13 | 1362 | |||
| AK170948.1 | - | 11, 12 | 1445 | |||
| AK171030.1 | - | 11, 12 | 1445 | |||
| AX775895.1 | - | 12, 13 | 1374 | |||
| AX775897.1 | - | 12, 13 | 1317 | |||
| BC016104.1 | - | 12, 13 | 1343 | 4920840 | ||

Gpr108
Genomic Map