This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C78339 SIRNA Assays ›
Gene Symbol:
C78339
Gene Name:
expressed sequence C78339
Gene Aliases:
-
Chromosome Location:
Chr.13: 46669443 - 46678064 on Build GRCm38
UniGene ID:
Mm.275934
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS294771
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C78339
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033192.2 NP_001028364.1 2 787
GenBank mRNA AK090371.1 - 2 171
AK141282.1 - 2 788
AK171039.1 - 3 688
AK218359.1 - 1 66

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_016255, BC047881, AK312814, AF097027, AK296393, and others.
This siRNA also targets the following transcripts in Rat: XM_008771560, XM_214446.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-8bmkk:80/100.66.79.215:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0