siRNA Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053723.1 | NP_446175.1 | 9 | 1323 | ||
| GenBank mRNA | AJ131111.1 | - | 9 | 1323 | ||
| BC085811.1 | - | 9 | 1301 | 7115423 | ||
| FQ211687.1 | - | 4 | 428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lancl1| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053723.1 | NP_446175.1 | 9 | 1323 | ||
| GenBank mRNA | AJ131111.1 | - | 9 | 1323 | ||
| BC085811.1 | - | 9 | 1301 | 7115423 | ||
| FQ211687.1 | - | 4 | 428 | |||