siRNA Details
| Gene Symbol: Il1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053953.1 | NP_446405.1 | 9 | 1265 | ||
| XM_008766983.1 | XP_008765205.1 | 9 | 1650 | |||
| XM_008766984.1 | XP_008765206.1 | 9 | 1421 | |||
| GenBank mRNA | BC091564.1 | - | 9 | 1239 | 7445971 | |
| FQ228742.1 | - | 9 | 1264 | |||
| FQ229102.1 | - | 9 | 1262 | |||
| Z22812.1 | - | 9 | 1265 | |||

Il1r2
Genomic Map