This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PDE11A SIRNA Assays ›
Gene Symbol:
Pde11a
Gene Name:
phosphodiesterase 11A
Gene Aliases:
-
Chromosome Location:
Chr.3: 62818502 - 63211843 on Build Rnor_6.0
UniGene ID:
Rn.88630
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS301086
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pde11a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127480.1 NP_001120952.1 11 1163
NM_001127481.2 NP_001120953.1 9 767
NM_080893.1 NP_543169.1 9 1841
GenBank mRNA AB059360.1 - 9 769
AB059361.1 - 11 1163
AB059362.1 - 9 1841

Back To Top

More Information


Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

cAMP catabolic process
signal transduction
cGMP catabolic process
cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
cAMP binding
cGMP binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0