siRNA Details
| Gene Symbol: Ythdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133423.1 | NP_596914.1 | 11 | 1498 | ||
| XM_006250805.2 | XP_006250867.1 | 12 | 1850 | |||
| XM_006250806.2 | XP_006250868.1 | 11 | 1813 | |||
| XM_006250808.2 | XP_006250870.1 | 10 | 1759 | |||
| XM_008769987.1 | XP_008768209.1 | 11 | 2865 | |||
| XM_008769988.1 | XP_008768210.1 | 11 | 2865 | |||
| XM_008769989.1 | XP_008768211.1 | 10 | 2811 | |||
| XM_008769990.1 | XP_008768212.1 | 11 | 2866 | |||
| GenBank mRNA | AF144731.1 | - | 11 | 1498 | ||
| D78303.1 | - | 10 | 1760 | |||

Ythdc1
Genomic Map