siRNA Details
| Gene Symbol: Casr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016996.1 | NP_058692.1 | 7 | 2843 | ||
| GenBank mRNA | AY214122.1 | - | 7 | 2833 | ||
| U10354.1 | - | 7 | 2843 | |||
| U20289.1 | - | 6 | 2469 | |||
More Information
Set Membership: |
|

Casr
Genomic Map