siRNA Details
| Gene Symbol: Tpcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139332.3 | NP_647548.2 | 454 | |||
| GenBank mRNA | AB018253.2 | - | 457 | |||
| BC062072.1 | - | 454 | 5599545 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpcn1| Gene Symbol: Tpcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139332.3 | NP_647548.2 | 454 | |||
| GenBank mRNA | AB018253.2 | - | 457 | |||
| BC062072.1 | - | 454 | 5599545 | |||