siRNA Details
| Gene Symbol: Fyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012755.1 | NP_036887.1 | 10, 11 | 1277 | ||
| XM_006256520.2 | XP_006256582.1 | 8, 9 | 2112 | |||
| XM_006256521.2 | XP_006256583.1 | 8, 9 | 2103 | |||
| XM_006256522.1 | XP_006256584.1 | 9, 10 | 1564 | |||
| GenBank mRNA | FQ231621.1 | - | 1, 2 | 162 | ||
| FQ232093.1 | - | 3, 4 | 377 | |||
| U35365.1 | - | 10, 11 | 1277 | |||

Fyn
Genomic Map