siRNA Details
| Gene Symbol: Slc38a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145776.1 | NP_665719.1 | 13 | 1168 | ||
| XM_006243708.2 | XP_006243770.1 | 13 | 1147 | |||
| GenBank mRNA | AF273025.1 | - | 13 | 1168 | ||
| BC081717.1 | - | 13 | 1189 | 7127614 | ||

Slc38a3
Genomic Map