This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ITGB7 SIRNA Assays ›
Gene Symbol:
Itgb7
Gene Name:
integrin, beta 7
Gene Aliases:
-
Chromosome Location:
Chr.7: 143820236 - 143837806 on Build Rnor_6.0
UniGene ID:
Rn.10767
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS304097
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Itgb7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013171.1 NP_037303.1 4 618
XM_006242383.2 XP_006242445.1 5 714
GenBank mRNA AF003598.1 - 2 117

Back To Top

More Information


Panther Classification:

Molecular Function -

integrin

Gene Ontology Categories:

Function(s) Process(es)

cell-matrix adhesion involved in ameboidal cell migration
cell-matrix adhesion
integrin-mediated signaling pathway
multicellular organism development
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
receptor clustering
leukocyte migration
leukocyte tethering or rolling
T cell migration
receptor activity
cell adhesion molecule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline