siRNA Details
| Gene Symbol: Tubd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105826.1 | NP_001099296.1 | 5 | 711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tubd1| Gene Symbol: Tubd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105826.1 | NP_001099296.1 | 5 | 711 | ||