siRNA Details
| Gene Symbol: Eme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105830.2 | NP_001099300.2 | 5, 6 | 1238 | ||
| XM_006247179.2 | XP_006247241.1 | 6, 7 | 1307 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eme1| Gene Symbol: Eme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105830.2 | NP_001099300.2 | 5, 6 | 1238 | ||
| XM_006247179.2 | XP_006247241.1 | 6, 7 | 1307 | |||