siRNA Details
| Gene Symbol: Adamts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137622.1 | NP_001131094.1 | 10 | 1629 | ||
| XM_008767677.1 | XP_008765899.1 | 8 | 1460 | |||
| XM_008767678.1 | XP_008765900.1 | 8 | 1460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts2| Gene Symbol: Adamts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137622.1 | NP_001131094.1 | 10 | 1629 | ||
| XM_008767677.1 | XP_008765899.1 | 8 | 1460 | |||
| XM_008767678.1 | XP_008765900.1 | 8 | 1460 | |||