siRNA Details
| Gene Symbol: Usp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100501.1 | NP_001093971.1 | 17 | 2359 | ||
| XM_006248015.2 | XP_006248077.1 | 17 | 2375 | |||
| GenBank mRNA | BC087705.1 | - | 16 | 2223 | ||
| BI282215.1 | - | 2 | 541 | |||
| FQ221582.1 | - | 16 | 3230 | |||
| FQ226796.1 | - | 4 | 825 | |||

Usp16
Genomic Map