siRNA Details
| Gene Symbol: Usp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100501.1 | NP_001093971.1 | 14 | 1804 | ||
| XM_006248015.2 | XP_006248077.1 | 14 | 1820 | |||
| GenBank mRNA | BC087705.1 | - | 13 | 1668 | ||
| BC112386.1 | - | 14 | 1754 | |||
| FQ221582.1 | - | 14 | 2770 | |||
| FQ226796.1 | - | 1 | 270 | |||

Usp16
Genomic Map