siRNA Details
| Gene Symbol: Hspa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100658.2 | NP_001094128.2 | 2, 3 | 244 | ||
| GenBank mRNA | S75280.1 | - | 2, 3 | 191 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa9| Gene Symbol: Hspa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100658.2 | NP_001094128.2 | 2, 3 | 244 | ||
| GenBank mRNA | S75280.1 | - | 2, 3 | 191 | ||