siRNA Details
| Gene Symbol: Ctu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037094.1 | NP_001032171.1 | 5 | 330 | ||
| GenBank mRNA | BC079393.1 | - | 5 | 380 | ||
| BC089862.1 | - | 5 | 328 | |||
| BC107464.1 | - | 5 | 330 | 7454431 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctu2| Gene Symbol: Ctu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037094.1 | NP_001032171.1 | 5 | 330 | ||
| GenBank mRNA | BC079393.1 | - | 5 | 380 | ||
| BC089862.1 | - | 5 | 328 | |||
| BC107464.1 | - | 5 | 330 | 7454431 | ||