siRNA Details
| Gene Symbol: RGD1305235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017447.1 | NP_001017447.1 | 2, 3 | 222 | ||
| GenBank mRNA | BC078920.1 | - | 2, 3 | 222 | 7096641 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305235| Gene Symbol: RGD1305235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017447.1 | NP_001017447.1 | 2, 3 | 222 | ||
| GenBank mRNA | BC078920.1 | - | 2, 3 | 222 | 7096641 | |