This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BCLAF1 SIRNA Assays ›
Gene Symbol:
Bclaf1
Gene Name:
BCL2-associated transcription factor 1
Gene Aliases:
Aa2-041
Chromosome Location:
Chr.1: 15799753 - 15828838 on Build Rnor_6.0
UniGene ID:
Rn.15652
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS308063
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bclaf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047852.2 NP_001041317.2 3 246
XM_006227652.2 XP_006227714.1 4 413
XM_006227653.2 XP_006227715.1 2 242
XM_006227654.2 XP_006227716.1 3 276
XM_006227655.2 XP_006227717.1 3 276
XM_006227656.2 XP_006227718.1 3 276
XM_008758605.1 XP_008756827.1 3 1405
GenBank mRNA BC161968.1 - 3 246 9028507

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001025393, NM_001025392, NM_153787, AK161692, AK150017, and others.

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of apoptotic process
regulation of DNA-templated transcription in response to stress
negative regulation of transcription, DNA-templated
positive regulation of DNA-templated transcription, initiation
positive regulation of response to DNA damage stimulus
positive regulation of intrinsic apoptotic signaling pathway
DNA binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-82hgg:80/100.66.79.225:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0