This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RGD735065 SIRNA Assays ›
Gene Symbol:
RGD735065
Gene Name:
similar to GI:13385412-like protein splice form I
Gene Aliases:
-
Chromosome Location:
Chr.20: 6869753 - 6910025 on Build Rnor_6.0
UniGene ID:
Rn.234381
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS309177
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD735065
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199379.2 NP_955411.1 6 712
XM_006256172.2 XP_006256234.1 5 1127
XM_006256173.2 XP_006256235.1 5 737
XM_006256174.2 XP_006256236.1 6 826
XM_006256175.2 XP_006256237.1 6 960
XM_006256176.2 XP_006256238.1 7 921
XM_006256177.2 XP_006256239.1 6 1217
XM_006256178.2 XP_006256240.1 7 1312
XM_006256180.2 XP_006256242.1 6 1267
XM_006256181.2 XP_006256243.1 5 1125
XM_006256182.2 XP_006256244.1 5 1121
XM_008772724.1 XP_008770946.1 6 1241
XM_008772725.1 XP_008770947.1 5 868
GenBank mRNA BC062242.1 - 6 713 6888059
FQ213349.1 - 3 328
FQ226355.1 - 3 328

Back To Top

More Information


Gene Ontology Categories:

Function(s)

wound healing
positive regulation of cell cycle
epithelial cell proliferation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-47mq8:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline