siRNA Details
| Gene Symbol: Eif3m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168523.1 | NP_001161995.1 | 6 | 632 | ||
| GenBank mRNA | FQ220907.1 | - | 6 | 605 | ||
| FQ222771.1 | - | 6 | 624 | |||
| FQ229546.1 | - | 6 | 625 | |||
| FQ229811.1 | - | 6 | 626 | |||

Eif3m
Genomic Map