siRNA Details
| Gene Symbol: LOC299271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181630.2 | NP_853661.1 | 4 | 1175 | ||
| GenBank mRNA | AY113703.1 | - | 4 | 1175 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC299271| Gene Symbol: LOC299271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181630.2 | NP_853661.1 | 4 | 1175 | ||
| GenBank mRNA | AY113703.1 | - | 4 | 1175 | ||