This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SETD3 SIRNA Assays ›
Gene Symbol:
Setd3 LOC100910833
Gene Name:
SET domain containing 3
histone-lysine N-methyltransferase setd3-like
Gene Aliases:
RGD1309550
-
Chromosome Location:
Chr.6: 132023128 - 132090070 on Build Rnor_6.0
Chromosome Location:
Chr.6: 131602143 - 131650422 on Build Rnor_6.0
UniGene ID:
Rn.7951
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS312191
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Setd3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_216781.8 XP_216781.6 6 782
GenBank mRNA BC089108.1 - 1 182
Gene Symbol: LOC100910833
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764908.1 XP_008763130.1 3 2185

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028262, AK166570, AK011993, BC019973, AY513271, and others.

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

histone H3-K36 methylation
peptidyl-lysine methylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of muscle cell differentiation
histone H3-K4 methylation
regulation of nucleic acid-templated transcription
RNA polymerase II activating transcription factor binding
transcription coactivator activity
histone methyltransferase activity (H3-K4 specific)
histone methyltransferase activity (H3-K36 specific)

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0