This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLR1314 SIRNA Assays ›
Gene Symbol:
Olr1314 LOC103693060
Gene Name:
olfactory receptor 1314
olfactory receptor 148-like
Gene Aliases:
-
-
Chromosome Location:
Chr.8: 43442614 - 43443531 on Build Rnor_6.0
Chromosome Location:
Chr.8: 43496457 - 43497374 on Build Rnor_6.0
UniGene ID:
Rn.143483
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS312812
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr1314
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000468.1 NP_001000468.1 559
Gene Symbol: LOC103693060
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766126.1 XP_008764348.1 1 559

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-6mqgs:80/100.66.73.142:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline