siRNA Details
| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191587.1 | NP_001178516.1 | 3 | 3220 | ||
| XM_006257523.2 | XP_006257585.1 | 3 | 4694 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcorl1| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191587.1 | NP_001178516.1 | 3 | 3220 | ||
| XM_006257523.2 | XP_006257585.1 | 3 | 4694 | |||