siRNA Details
| Gene Symbol: Fscn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107072.1 | NP_001100542.1 | 1 | 249 | ||
| XM_008768476.1 | XP_008766698.1 | 1 | 5406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fscn2| Gene Symbol: Fscn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107072.1 | NP_001100542.1 | 1 | 249 | ||
| XM_008768476.1 | XP_008766698.1 | 1 | 5406 | |||