siRNA Details
| Gene Symbol: Il10rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107111.1 | NP_001100581.1 | 3 | 275 | ||
| XM_006248062.2 | XP_006248124.1 | 3 | 504 | |||
| XM_006248063.2 | XP_006248125.1 | 3 | 848 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il10rb| Gene Symbol: Il10rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107111.1 | NP_001100581.1 | 3 | 275 | ||
| XM_006248062.2 | XP_006248124.1 | 3 | 504 | |||
| XM_006248063.2 | XP_006248125.1 | 3 | 848 | |||