siRNA Details
| Gene Symbol: Adamts5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198761.1 | NP_942056.1 | 3, 4 | 1396 | ||
| GenBank mRNA | AY382879.1 | - | 3, 4 | 1396 | ||
| FQ228283.1 | - | 1789 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts5| Gene Symbol: Adamts5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198761.1 | NP_942056.1 | 3, 4 | 1396 | ||
| GenBank mRNA | AY382879.1 | - | 3, 4 | 1396 | ||
| FQ228283.1 | - | 1789 | ||||