siRNA Details
| Gene Symbol: Cers4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107117.1 | NP_001100587.1 | 7 | 678 | ||
| XM_006248804.2 | XP_006248866.1 | 6 | 794 | |||
| XM_006248805.2 | XP_006248867.1 | 6 | 696 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cers4| Gene Symbol: Cers4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107117.1 | NP_001100587.1 | 7 | 678 | ||
| XM_006248804.2 | XP_006248866.1 | 6 | 794 | |||
| XM_006248805.2 | XP_006248867.1 | 6 | 696 | |||