siRNA Details
| Gene Symbol: Zfp648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769689.1 | XP_008767911.1 | 1 | 410 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp648| Gene Symbol: Zfp648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769689.1 | XP_008767911.1 | 1 | 410 | ||