This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DSP SIRNA Assays ›
Gene Symbol:
Dsp
Gene Name:
desmoplakin
Gene Aliases:
-
Chromosome Location:
Chr.17: 27286811 - 27334451 on Build Rnor_6.0
UniGene ID:
Rn.231913
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS316453
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dsp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_225259.8 XP_225259.4 18 2877

Back To Top

More Information


Panther Classification:

Molecular Function -

intermediate filament binding protein

Gene Ontology Categories:

Function(s) Process(es)

desmosome organization
ventricular compact myocardium morphogenesis
single organismal cell-cell adhesion
peptide cross-linking
keratinocyte differentiation
adherens junction organization
wound healing
skin development
intermediate filament cytoskeleton organization
intermediate filament organization
protein localization to adherens junction
ventricular cardiac muscle cell action potential
bundle of His cell to Purkinje myocyte communication
regulation of heart rate by cardiac conduction
protein kinase C binding
structural molecule activity
protein binding, bridging
poly(A) RNA binding
cell adhesion molecule binding
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0