siRNA Details
| Gene Symbol: Smpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006997.1 | NP_001006998.1 | 2 | 459 | ||
| GenBank mRNA | BC083780.1 | - | 2 | 459 | 7096039 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smpd1| Gene Symbol: Smpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006997.1 | NP_001006998.1 | 2 | 459 | ||
| GenBank mRNA | BC083780.1 | - | 2 | 459 | 7096039 | |