siRNA Details
| Gene Symbol: Wbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127484.1 | NP_001120956.1 | 4 | 471 | ||
| XM_006231508.2 | XP_006231570.1 | 4 | 2466 | |||
| XM_008760435.1 | XP_008758657.1 | 4 | 1301 | |||
| XM_008760436.1 | XP_008758658.1 | 4 | 2578 | |||
| GenBank mRNA | BC166754.1 | - | 4 | 408 | 7929564 | |
| FQ217301.1 | - | 4 | 716 | |||

Wbp1l
Genomic Map