This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC35E2B SIRNA Assays ›
Gene Symbol:
Slc35e2b
Gene Name:
solute carrier family 35, member E2B
Gene Aliases:
Slc35e2
Chromosome Location:
Chr.5: 173024335 - 173050228 on Build Rnor_6.0
UniGene ID:
Rn.158991
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS320458
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc35e2b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107998.1 NP_001101468.1 4 1458
XM_006239572.2 XP_006239634.1 4 1020
XM_006239573.2 XP_006239635.1 4 1137
XM_008764361.1 XP_008762583.1 4 1137

Back To Top

More Information


Panther Classification:

Molecular Function -

secondary carrier transporter

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline