siRNA Details
| Gene Symbol: Asxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764542.1 | XP_008762764.1 | 12 | 3166 | ||
| XM_008764543.1 | XP_008762765.1 | 11 | 3022 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asxl2| Gene Symbol: Asxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764542.1 | XP_008762764.1 | 12 | 3166 | ||
| XM_008764543.1 | XP_008762765.1 | 11 | 3022 | |||