siRNA Details
| Gene Symbol: Bcl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277287.1 | NP_001264216.1 | 4 | 855 | ||
| NM_001277288.1 | NP_001264217.1 | 3 | 651 | |||
| XM_006240540.2 | XP_006240602.1 | 3 | 814 | |||
| GenBank mRNA | JF520428.1 | - | 3 | 651 | ||
| JF520429.1 | - | 4 | 855 | |||

Bcl11b
Genomic Map