siRNA Details
| Gene Symbol: Espl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170602.1 | NP_001164073.1 | 18 | 3956 | ||
| XM_006242460.2 | XP_006242522.1 | 18 | 4101 | |||
| XM_006242461.2 | XP_006242523.1 | 18 | 4096 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Espl1| Gene Symbol: Espl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170602.1 | NP_001164073.1 | 18 | 3956 | ||
| XM_006242460.2 | XP_006242522.1 | 18 | 4101 | |||
| XM_006242461.2 | XP_006242523.1 | 18 | 4096 | |||