siRNA Details
| Gene Symbol: Csk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030039.1 | NP_001025210.1 | 8, 9 | 871 | ||
| XM_006243163.1 | XP_006243225.1 | 9, 10 | 1517 | |||
| XM_006243164.2 | XP_006243226.1 | 9, 10 | 1200 | |||
| GenBank mRNA | BC098863.1 | - | 8, 9 | 871 | 7380519 | |
| X58631.1 | - | 9, 10 | 906 | |||

Csk
Genomic Map