siRNA Details
| Gene Symbol: Dzip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014095.1 | NP_001014117.1 | 6 | 1044 | ||
| XM_006243626.2 | XP_006243688.1 | 7 | 1248 | |||
| XM_006243627.2 | XP_006243689.1 | 7 | 1399 | |||
| XM_008766526.1 | XP_008764748.1 | 6 | 2012 | |||
| XM_008766527.1 | XP_008764749.1 | 6 | 2012 | |||
| GenBank mRNA | BC083786.1 | - | 6 | 1044 | 7097259 | |

Dzip1l
Genomic Map