siRNA Details
| Gene Symbol: Plin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_236783.6 | XP_236783.4 | 2 | 147 | ||
| GenBank mRNA | FQ230643.1 | - | 2 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plin3| Gene Symbol: Plin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_236783.6 | XP_236783.4 | 2 | 147 | ||
| GenBank mRNA | FQ230643.1 | - | 2 | 84 | ||