siRNA Details
| Gene Symbol: Tbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181638.1 | NP_853669.1 | 1 | 15 | ||
| XM_006249415.2 | XP_006249477.1 | 2 | 1163 | |||
| XM_006249416.2 | XP_006249478.1 | 2 | 2026 | |||
| XM_006249417.2 | XP_006249479.1 | 2 | 1173 | |||
| XM_006249418.1 | XP_006249480.1 | 1 | 1287 | |||
| GenBank mRNA | AY289619.1 | - | 1 | 15 | ||
| FQ221880.1 | - | 1 | 985 | |||

Tbx3
Genomic Map