siRNA Details
| Gene Symbol: Gramd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191826.1 | NP_001178755.1 | 495 | |||
| XM_008768766.1 | XP_008766988.1 | 6 | 653 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gramd1c| Gene Symbol: Gramd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191826.1 | NP_001178755.1 | 495 | |||
| XM_008768766.1 | XP_008766988.1 | 6 | 653 | |||