siRNA Details
Gene Symbol: Suco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199403.1 | NP_955435.1 | 15, 16 | 1500 | ||
XM_006250164.2 | XP_006250226.1 | 15, 16 | 1955 | |||
XM_006250166.2 | XP_006250228.1 | 14, 15 | 1934 | |||
XM_008769657.1 | XP_008767879.1 | 15, 16 | 1967 | |||
XM_008769658.1 | XP_008767880.1 | 14, 15 | 1946 | |||
XM_008769659.1 | XP_008767881.1 | 15, 16 | 1967 | |||
GenBank mRNA | AJ421447.1 | - | 15, 16 | 1500 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172645, BC138379, AK029097, AK171856.
Panther Classification:
Gene Ontology Categories:
