siRNA Details
| Gene Symbol: Gtdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024274.1 | NP_001019445.1 | 555 | |||
| GenBank mRNA | AY281367.1 | - | 555 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtdc1| Gene Symbol: Gtdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024274.1 | NP_001019445.1 | 555 | |||
| GenBank mRNA | AY281367.1 | - | 555 | |||